Learn More
IDEX Health & Science Upchurch STAINLESS STEEL FRIT 0.5
Catalog No. NC1298747
Encompass_Preferred
Stainless Steel Frit 0.5µm .094" x .065" x .250" Single
Catalog No. NC1298747 Supplier IDEX Health & Science Upchurch Supplier No. A103
Stainless Steel Frit 0.5µm .094" x .065" x .250" Single