Learn More
Cole-Parmer Cole-Parmer Essentials Stainless Steel Funnel, 32 oz
Catalog No. NC1570519 Shop All Cole-Parmer Products
Encompass_Preferred
Cole-Parmer Stainless Steel Funnel, 32 oz, Cole-Parmer stainless steel funnels are a laboratory supply you will count on for years of use. Stay in stock with our selection of medium gauge, 304 stainless steel funnels in various capacities and overall sizes.
Catalog No. NC1570519 Supplier Cole-Parmer Supplier No. 0726700
Cole-Parmer Stainless Steel Funnel, 32 oz, Cole-Parmer stainless steel funnels are a laboratory supply you will count on for years of use. Stay in stock with our selection of medium gauge, 304 stainless steel funnels in various capacities and overall sizes.