Learn More
Perkin Elmer US LLC Stainless Steel GSV Union, 0.0625 in I.D.
Catalog No. 502112508
Encompass_Preferred
Used with Arnel Analyzers. Bore: 0.029 inch (0.75 mm) Hex: 1/4 inch Tubing Size: 1/16 inch
Catalog No. 50-211-2508 Supplier Perkin Elmer US LLC Supplier No. N9302839
Used with Arnel Analyzers. Bore: 0.029 inch (0.75 mm) Hex: 1/4 inch Tubing Size: 1/16 inch