Learn More
TA Instruments Stainless Steel Helical Rotor for HR X0, DHR and AR-G2, Diameter 30 mm, Length 31 mm, Requires Helical Cup P/N 546093.001 and Smart Swap Concentric Cylinder Peltier Jacket P/N 533201.901
Catalog No. 5000580699 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Stainless Steel Helical Rotor For HR X0, DHR and AR-G2 has a diameter of 30 mm and length of 31 mm. It requires the Helical Cup (P/N 546093.001) and HR X0, DHR, and AR-Series Smart Swap Concentric Cylinder Peltier Jacket (P/N 533201.901).
Catalog No. 50-005-80699 Supplier TA Instruments Supplier No. 546018.901
Waters TA Instruments Stainless Steel Helical Rotor For HR X0, DHR and AR-G2 has a diameter of 30 mm and length of 31 mm. It requires the Helical Cup (P/N 546093.001) and HR X0, DHR, and AR-Series Smart Swap Concentric Cylinder Peltier Jacket (P/N 533201.901).