Learn More
Cole-Parmer Troemner 12310-ST 2 kg, Class F Stainless Steel Hook Top Weight with Traceable Certificate
Catalog No. NC1708095 Shop All Cole-Parmer Products
Encompass_Preferred
Troemner’s Stainless Steel Hook weights are manufactured from 300 series stainless steel. Each weight has the nominal value stamped into the top side of the weight. Hook Top Only - Hook Top Weights are flat on top with a stationary hook in the center of the weight
Catalog No. NC1708095 Supplier Cole-Parmer Supplier No. 0103422
Troemner’s Stainless Steel Hook weights are manufactured from 300 series stainless steel. Each weight has the nominal value stamped into the top side of the weight. Hook Top Only - Hook Top Weights are flat on top with a stationary hook in the center of the weight