Learn More
Medicus Health IV Poles - Stainless Steel with Hand Crank, 8 Hooks
SDP
Catalog No. NC2541531
Encompass
∙ IV Pole with 8 Hooks have heavy-duty turn crank easily lift heavy loads and help reduce back injuries IV Poles with heavy-duty turn crank easily lift heavy loads∙ Helps reduce back injuries∙ Height-adjustable from 56" - 96"∙ Six leg base offer supreme stability∙ Pole weighs 55 lbs., ships assembled and must travel via freight carrier∙ 3" scuff resistant casters provide unmatched maneuverability∙ Base post is 2" square stainless steel∙ Extension post is 1.25" stainless steel tube∙ IV Pole weight capacity is approximately 60 lbs.∙ No electric motors or hydraulic pumps to be concerned about or replaced
This item is not returnable. View return policy
Catalog No. NC2541531 Supplier Medicus Health Supplier No. 2238M
∙ IV Pole with 8 Hooks have heavy-duty turn crank easily lift heavy loads and help reduce back injuries IV Poles with heavy-duty turn crank easily lift heavy loads∙ Helps reduce back injuries∙ Height-adjustable from 56" - 96"∙ Six leg base offer supreme stability∙ Pole weighs 55 lbs., ships assembled and must travel via freight carrier∙ 3" scuff resistant casters provide unmatched maneuverability∙ Base post is 2" square stainless steel∙ Extension post is 1.25" stainless steel tube∙ IV Pole weight capacity is approximately 60 lbs.∙ No electric motors or hydraulic pumps to be concerned about or replaced