Learn More
Justrite™ Stainless Steel Joining Clips for Connecting EcoPolyBlend™ Accumulation Centers
Catalog No. 19822466 Shop All Justrite Safety Group Products
Change view
Click to view available options
Material:
Stainless Steel
Catalog No. Material
19-822-466 Stainless Steel
1 options
Catalog No. 19-822-466 Supplier Justrite™ Supplier No. 28926
Add to Cart
Add to Cart
Bring old equipment back into service with replacement parts. The Justrite™ Stainless Steel Joining Clips for Connecting EcoPolyBlend™ Accumulation Centers is an accessory to help facilitate the use of accumulation centers.
By joining centers together with these clips or the patented Sump-to-Sump Drain™ kit for a total 66 gal. sump capacity, you will meet EPA and UFC requirements.
For compliant storage using unites 4-drum and smaller, use these joining clips to reach needed capacity. Also available as a kit (19-822-446).
Specifications
Color Silver
Material Stainless Steel
Width (English) 2 in.
Height (English) 4 in.