Learn More
Chemglass Life Sciences Stainless Steel Keck Style Clamp, #24
SDP
Catalog No. 501531110
Encompass_Preferred
Stainless Steel Keck Style Clamp, #24. Stainless steel clamps to fit 24/40 and 24/25 standard taper joints. Metal clamps are suitable for high temperatures applications.
Catalog No. 50-153-1110 Supplier Chemglass Life Sciences Supplier No. CG145M24
Stainless Steel Keck Style Clamp, #24. Stainless steel clamps to fit 24/40 and 24/25 standard taper joints. Metal clamps are suitable for high temperatures applications.