Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Stainless Steel Laboratory Bottle, with DIN 168-1 Thread, GL 45
SDP

Catalog No. 02900136 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
1500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
02-900-136 1500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-900-136 Supplier DWK Life Sciences Supplier No. 299016006
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Stainless Steel Laboratory Bottle, with DIN 168-1 Thread, GL 45 is ideal for storage where glass is unsuitable. Made from corrosion-resistant AISI 316L stainless steel with a smooth inner finish, built-in pouring lip, and full GL 45 cap compatibility.

  • Manufactured from corrosion-resistant AISI Type 316L (1.4404) stainless steel with hygienically welded seams
  • Smooth inner surface finish corresponding to IIIc (DIN 17441) with rounded inner edges for easy cleaning
  • Polished and brushed durable exterior finish
  • GL 45 thread with built-in pouring lip for clean, controlled pouring
  • Fully compatible with all GL 45 caps; stainless steel cap with PTFE sealing disc available separately
  • Standardised GL thread and matching cap system for particularly tight sealing
  • Ideal for storage applications where glass is not suitable due to breakage risk or chemical incompatibility
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 1500 mL
Color Silver
For Use With (Application) Storage, transport and safe handling of hazardous, dangerous and valuable materials; Storage and transport of light-sensitive materials
Material Stainless Steel (316L)
Closure Included No
Capacity (English) 51 oz.
Shape Round
Product Type Storage Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.