Learn More
Arrow Engineering S-316-30 Type 316 Stainless Steel 30" Propeller Shaft
Catalog No. NC2226902
Encompass_Preferred
All Arrow Mixing Products' stainless steel mixer shafts are 3/8" dia. & made of 316 stainless steel. These quality mixer parts have a 5/16-24 external thread for easy assembly with all of our stainless steel propellers and couplings. (Note: propeller and coupling sold separately)
This item is not returnable. View return policy
Catalog No. NC2226902 Supplier Arrow Engineering Supplier No. S31630
All Arrow Mixing Products' stainless steel mixer shafts are 3/8" dia. & made of 316 stainless steel. These quality mixer parts have a 5/16-24 external thread for easy assembly with all of our stainless steel propellers and couplings. (Note: propeller and coupling sold separately)