Learn More
AVANTOR MASTERFLEX (VWR LLC) Stainless steel mounting hardware; for one L/S Easy-Load II pump head.
Catalog No. NC1006472
Encompass
Masterflex Stainless steel mounting hardware; for one L/S Easy-Load II pump head. Designed to make the Masterflex L/S series pump head flexible for a wide range of applicationsChoose from different material or increase the number of pump heads on your L/S drive with mounting hardwareCompatible with the Masterflex L/S series peristaltic drives
Catalog No. NC1006472 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX7720001
Masterflex Stainless steel mounting hardware; for one L/S Easy-Load II pump head. Designed to make the Masterflex L/S series pump head flexible for a wide range of applicationsChoose from different material or increase the number of pump heads on your L/S drive with mounting hardwareCompatible with the Masterflex L/S series peristaltic drives