Learn More
Contec™ Stainless Steel Multi-Bucket System
Catalog No. 19161020 Shop All Contec Inc. Products
Change view
Click to view available options
Product Type:
Multiple Bucket System
Catalog No. Product Type
19-161-020 Multiple Bucket System
1 options
Catalog No. 19-161-020 Supplier Contec™ Supplier No. 2707
Stainless Steel Multi-Bucket System offers maximum flexibility and can be used with two or three 7 gallon (27 L) buckets depending upon your in-house protocols.
  • Lightweight, stainless steel, and easy to maneuver multi-bucket system offers maximum flexibility and can be used with two or three 7- gallon (27 L) buckets depending upon in-house protocols.
  • An extendable frame pulls out of the front of the fully autoclavable cart to accommodate an additional bucket, part number 2617.
  • The cart features non-marring run quiet wheels.
  • Buckets are deep drawn and seamless reducing the risk of microbial contamination
  • The removable handle makes it convenient for autoclaving and storage.
  • A lip around the top edge of the buckets allows for easy lifting.
  • Autoclavable at 121°C for 30 minutes
  • 2700 multi-bucket system includes heavy-duty stainless steel downpress wringer that is compatible with all sizes of VertiKlean™, VertiKlean™ MAX™, QuickTask™, and Edgeless™ Mops.
  • 2707 multi-bucket system offers the ability to customize the system with whichever wringer best coincides with in-house protocols. The system is compatible with two stainless steel downpress wringer options to accommodate both flat and string mops
Specifications
Product Type Multiple Bucket System
Material Stainless Steel
Autoclavable Autoclavable
Disposable Reusable
For Use With (Application) Ideal for use in life sciences, microelectronics, semiconductor or any critical manufacturing environment
Includes Two 7 gal. Buckets, cart with casters (wringer options sold separately)
Sterility Non-sterile