Learn More
Perkin Elmer US LLC Stainless Steel Nebulizer Body for AAnalyst
Catalog No. 502112769
Encompass_Preferred
This stainless steel body is included in the AAnalyst Corrosion-Resistant Nebulizer Complete Assembly (03030404), Platinum Alloy Nebulizer Complete Assembly (03030299), and Stainless Steel Nebulizer Complete Assembly (03030352). This nebulizer body is compatible with the PerkinElmer AAnalyst 100/200/300/400/700/800 and Models 5100PC/5000/3300/3110/3100 AA instruments.
Catalog No. 50-211-2769 Supplier Perkin Elmer US LLC Supplier No. 03030353
This stainless steel body is included in the AAnalyst Corrosion-Resistant Nebulizer Complete Assembly (03030404), Platinum Alloy Nebulizer Complete Assembly (03030299), and Stainless Steel Nebulizer Complete Assembly (03030352). This nebulizer body is compatible with the PerkinElmer AAnalyst 100/200/300/400/700/800 and Models 5100PC/5000/3300/3110/3100 AA instruments.