Learn More
Perkin Elmer US LLC Stainless Steel Nebulizer End Cap Assembly with Viton O-Rings for PinAAcle
Catalog No. 502112115
Encompass_Preferred
This end cap assembly is included in the Standard Stainless Steel Nebulizer Complete Assembly for PinAAcle (N3160143) and the Organics Stainless Steel Nebulizer Complete Assembly for PinAAcle (N3160158). Note: This end cap ships as standard equipment on instruments configured with a stainless steel nebulizer.
Catalog No. 50-211-2115 Supplier Perkin Elmer US LLC Supplier No. N3160505
This end cap assembly is included in the Standard Stainless Steel Nebulizer Complete Assembly for PinAAcle (N3160143) and the Organics Stainless Steel Nebulizer Complete Assembly for PinAAcle (N3160158). Note: This end cap ships as standard equipment on instruments configured with a stainless steel nebulizer.