Learn More
Perkin Elmer US LLC Stainless Steel Nebulizer End Cap Assembly with Kalrez O-Rings for PinAAcle
Catalog No. 502104338
Encompass_Preferred
This end cap assembly is included in the Organics Stainless Steel Nebulizer Complete Assembly for PinAAcle (N3160188). Note: This end cap ships as standard equipment on instruments configured with a stainless steel nebulizer.
Catalog No. 50-210-4338 Supplier Perkin Elmer US LLC Supplier No. N3160188
This end cap assembly is included in the Organics Stainless Steel Nebulizer Complete Assembly for PinAAcle (N3160188). Note: This end cap ships as standard equipment on instruments configured with a stainless steel nebulizer.