Learn More
Perkin Elmer US LLC Stainless Steel Oxygen/Moisture Click-On Trap
Catalog No. 502104592
Encompass_Preferred
This moisture trap contains a spring loaded check valve seal when a filter is removed and open only when a new filter has been locked in. There is no need for loosening and tightening fittings every time a trap is changed and your system will remain uncontaminated during the process.
Catalog No. 50-210-4592 Supplier Perkin Elmer US LLC Supplier No. N9306103
This moisture trap contains a spring loaded check valve seal when a filter is removed and open only when a new filter has been locked in. There is no need for loosening and tightening fittings every time a trap is changed and your system will remain uncontaminated during the process.