Learn More
Perkin Elmer US LLC Stainless Steel Oxygen/Moisture Click-On Trap Kit
Catalog No. 502104617
Encompass_Preferred
Click-On connectors reduce system downtime and allow you to replace a trap without an open gas line, avoiding the introduction of impurities into the system. This in turn eliminates the need to flush the system. The ability to add a Click-On Inline Super Clean Indicator after the stainless steel trap gives the user a clear visual indication of when to change the filter. This indicator may also be used as a standalone trap. Connectors: 1/8 inch Brass Gas Quality: 100% Maximium Pressure: 11 bar, 160 psi Maximum Flow: 25 L/min Estimated Lifetime: >3 years
Catalog No. 50-210-4617 Supplier Perkin Elmer US LLC Supplier No. N9306108
Click-On connectors reduce system downtime and allow you to replace a trap without an open gas line, avoiding the introduction of impurities into the system. This in turn eliminates the need to flush the system. The ability to add a Click-On Inline Super Clean Indicator after the stainless steel trap gives the user a clear visual indication of when to change the filter. This indicator may also be used as a standalone trap. Connectors: 1/8 inch Brass Gas Quality: 100% Maximium Pressure: 11 bar, 160 psi Maximum Flow: 25 L/min Estimated Lifetime: >3 years