Learn More
Perkin Elmer US LLC Stainless Steel Pans, Covers and O-Rings - Qty. 1000 ea.
Catalog No. 502113480
Encompass_Preferred
These high pressure pans and covers contain a sample in a sealed environment throughout an experiment. This ensures that the heat generated from vaporization or a volatile reaction does not interfere with the analyzer's measurements. These pans and covers are compatible with the the DSC 4000/6000/8000/8500, DSC 6, DSC 7, Jade DSC, Diamond DSC, PYRIS 1 DSC, and PYRIS 6 DSC.
Catalog No. 50-211-3480 Supplier Perkin Elmer US LLC Supplier No. 03190029
Add to Cart
Add to Cart
Request a Quote
These high pressure pans and covers contain a sample in a sealed environment throughout an experiment. This ensures that the heat generated from vaporization or a volatile reaction does not interfere with the analyzer's measurements. These pans and covers are compatible with the the DSC 4000/6000/8000/8500, DSC 6, DSC 7, Jade DSC, Diamond DSC, PYRIS 1 DSC, and PYRIS 6 DSC.