Learn More
Perkin Elmer US LLC Stainless Steel Pans, Covers & O-Rings - Qty. 20 ea.
Catalog No. 502113483
Encompass_Preferred
These high pressure pans and covers contain a sample in a sealed environment throughout an experiment. This ensures that the heat generated from vaporization or a volatile reaction does not interfere with the analyzer's measurements. These pans are compatible with the DSC 6/7/4000/6000/8000/8500/Jade DSC/Diamond DSC/PYRIS 1 DSC/PYRIS 6 DSC.
Catalog No. 50-211-3483 Supplier Perkin Elmer US LLC Supplier No. 03190218
These high pressure pans and covers contain a sample in a sealed environment throughout an experiment. This ensures that the heat generated from vaporization or a volatile reaction does not interfere with the analyzer's measurements. These pans are compatible with the DSC 6/7/4000/6000/8000/8500/Jade DSC/Diamond DSC/PYRIS 1 DSC/PYRIS 6 DSC.