Learn More
Electron Microscopy Sciences Stainless Steel Probe #1
SDP
Catalog No. 50996770
Encompass_Preferred
These probes are scalpel sharp for fine probing. They are useful for many applications, such as micro positioning, solder probing, and band inspection.
Catalog No. 50-996-770 Supplier Electron Microscopy Sciences Supplier No. 6212511
These probes are scalpel sharp for fine probing. They are useful for many applications, such as micro positioning, solder probing, and band inspection.