Learn More
Electron Microscopy Sciences Stainless Steel Probe with Flat tip, 155 mm OAL
SDP
Catalog No. 501930651
Encompass_Preferred
Stainless steel probes are perfect for lab applications and electronics checking and inspections. The probes are made of rust free stainless steel and have a serrated handle for a better grip. Probe length is 155 mm.
OAL: 155 mm
Catalog No. 50-193-0651 Supplier Electron Microscopy Sciences Supplier No. 7832611
Stainless steel probes are perfect for lab applications and electronics checking and inspections. The probes are made of rust free stainless steel and have a serrated handle for a better grip. Probe length is 155 mm.
OAL: 155 mm