Learn More
Electron Microscopy Sciences Stainless Steel Probes, 3/Set
SDP
Catalog No. 50996769
Encompass_Preferred
These probes are scalpel sharp for fine probing. They are useful for many applications, such as micro positioning, solder probing, and band inspection.
Catalog No. 50-996-769 Supplier Electron Microscopy Sciences Supplier No. 6212510
These probes are scalpel sharp for fine probing. They are useful for many applications, such as micro positioning, solder probing, and band inspection.