Learn More
Micronova™ Stainless Steel Push Cart and Bucket System
SDP
Catalog No. 19121977 Shop All Micronova Manufacturing Products
Catalog No. 19-121-977 Supplier Micronova™ Supplier No. C227
A highly maneuverable, stainless steel, double bucket push cart system with an ergonomically formed push bar handle and Prevenz™ antimicrobial wheels with 360° rotation. System includes two rectangular 8 gallon (30 liter) smooth, molded stainless steel buckets.
The Micronova Stainless Steel Push Cart and Bucket System features an electropolished 304 stainless steel push cart with three grip-it mop holders located on the push bar for easy transport of mop handles and a foot activated brake for added stability when stopped. The included stainless steel buckets are molded with no ridges or seams to harbor bacteria.
  • Highly maneuverable, double bucket push cart system
  • Electropolished 304 stainless steel cart and buckets
  • Ergonomically formed push bar handle
  • Three grip-it mop holders located on the push bar
  • Prevenz™ antimicrobial wheels with 360° rotation
  • Foot activated brake for added stability when stopped
  • Durable bacteria-resistant construction
  • Rectangular 8 gallon (30 liter) buckets accept 18 inch wide mops
  • Compatible with Slim-T Wringer series
  • Entire system is autoclavable
    Specifications
    Autoclavable Autoclavable