Learn More
TA Instruments Stainless Steel Quick Change Plate, 4 mm, HR X0, DHR, ARES-G2, APS
Catalog No. 5000580639 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments HR X0, DHR, ARES-G2, APS Stainless Steel 4 mm Quick Change Plate is for use with HR X0/DHR Advanced Peltier Plate, High Temperature-Advanced Peltier Plate, and ARES-G2 Advanced Peltier System with the Standard Flat Plate.
Catalog No. 50-005-80639 Supplier TA Instruments Supplier No. 534595.901
Waters TA Instruments HR X0, DHR, ARES-G2, APS Stainless Steel 4 mm Quick Change Plate is for use with HR X0/DHR Advanced Peltier Plate, High Temperature-Advanced Peltier Plate, and ARES-G2 Advanced Peltier System with the Standard Flat Plate.