Learn More
Chemglass Life Sciences Stainless Steel Quick Disconnect Coupling Set: Coupler and Nipple with each side having a 3/4" Female NPT Fitting On Each End, Temp Range -60 to 200°C, Fluorosilicone
SDP
Supplier:  Chemglass Life Sciences CG1969M80
Encompass_Preferred
Stainless Steel Quick connects terminating in either both male or both female M30 thread. Quick connects have fluorosilicone seals. Both male and female couplers have shutoffs. Quick connects provide an easy and convenient way to change out reactor systems or disconnecting hoses. Temperature range is -60 to 200 degrees C. M30 fittings are compatible with the Chemglass ball valves, hoses and adapters. PLEASE NOTE: TWO QUICK CONNECT SETS ARE REQUIRED PER REACTOR SETUP. Special end fittings and configurations are available. Quick connects are not considered dripless/dry quick disconnect couplings.
Catalog No. 50-009-80681