Learn More
BINDER™ Rack
SDP
Catalog No. 08111844 Shop All BINDER Inc. Products
Change view
Click to view available options
Material:
Stainless Steel
Catalog No. Material
08-111-844 Stainless Steel
1 options
Catalog No. 08-111-844 Supplier BINDER™ Supplier No. 80122159
Add to Cart
Add to Cart
Material: Stainless Steel; For Use With: ED 400, FED 400, FP 400, M 400, BD 400, BF 400
  • Convenient storage space
  • Manufactured from stainless steel
Specifications
Description Rack - stainless steel
For Use With (Equipment) BINDER Chambers ED 400, FED 400, FP 400, M 400, BD 400, BF 400
Material Stainless Steel
Type Rack