Learn More
TA Instruments Stainless Steel, Sandblasted, 20 mm Plate, NST, HR X0
Catalog No. 5000580799 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments HR X0 Stainless Steel Sandblasted Upper Plate (20 mm, NST) is for use with lower Peltier plate. Smart Swap 2 geometry for use with HR X0 instruments only. Surface Finish: 80-84 RMS in micro inches or Ra value of 1.8 to 2.0 microns.
Catalog No. 50-005-80799 Supplier TA Instruments Supplier No. 519200.946
Waters TA Instruments HR X0 Stainless Steel Sandblasted Upper Plate (20 mm, NST) is for use with lower Peltier plate. Smart Swap 2 geometry for use with HR X0 instruments only. Surface Finish: 80-84 RMS in micro inches or Ra value of 1.8 to 2.0 microns.