Learn More
TA Instruments Stainless Steel, Sandblasted, 20 mm Plate, Smart Swap 2 Geometry, Surface Finish 80-84 RMS, HR x0 Instruments Only
Catalog No. 5000580798 Shop All Waters TA Instruments Products
Encompass_Preferred
HR X0 Stainless Steel Sandblasted Plate (20 mm) is an upper plate for use with lower Peltier plates. It features Smart Swap 2 geometry and a surface finish of 80-84 RMS (Ra 1.8 to 2.0 microns), ensuring reliable sample contact and consistent measurement for HR X0 instruments.
Catalog No. 50-005-80798 Supplier TA Instruments Supplier No. 519200.945
HR X0 Stainless Steel Sandblasted Plate (20 mm) is an upper plate for use with lower Peltier plates. It features Smart Swap 2 geometry and a surface finish of 80-84 RMS (Ra 1.8 to 2.0 microns), ensuring reliable sample contact and consistent measurement for HR X0 instruments.