Learn More
TA Instruments Stainless Steel, Sandblasted, 20 mm, Quick Change Plate, HR X0, DHR, ARES-G2, APS
Catalog No. 5000580725 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments HR X0, DHR, ARES-G2, APS Stainless Steel Sandblasted Quick Change Plate (20 mm) features a surface finish of 80-84 RMS (Ra 1.8 to 2.0 microns). Compatible with Advanced Peltier Plates and ARES-G2 APS systems for reliable alignment and reproducible measurements.
Catalog No. 50-005-80725 Supplier TA Instruments Supplier No. 402494.901
Waters TA Instruments HR X0, DHR, ARES-G2, APS Stainless Steel Sandblasted Quick Change Plate (20 mm) features a surface finish of 80-84 RMS (Ra 1.8 to 2.0 microns). Compatible with Advanced Peltier Plates and ARES-G2 APS systems for reliable alignment and reproducible measurements.