Learn More
TA Instruments Stainless Steel, Sandblasted, 40 mm Plate, Upper Plate for Use with Lower Peltier Plate, Smart Swap 2 Geometry, Surface Finish 80-84 RMS, Ra 1.8-2.0 Microns, HR x0 Instruments Only
Catalog No. 5000580835 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments HR X0, Stainless Steel, Sandblasted, 40 mm Plate is an upper plate for use with the lower Peltier plate. It has Smart Swap 2 geometry and is for use with HR X0 instruments only. Surface finish is 80-84 RMS in micro inches or Ra value of 1.8 to 2.0 microns.
Catalog No. 50-005-80835 Supplier TA Instruments Supplier No. 519400.945
Waters TA Instruments HR X0, Stainless Steel, Sandblasted, 40 mm Plate is an upper plate for use with the lower Peltier plate. It has Smart Swap 2 geometry and is for use with HR X0 instruments only. Surface finish is 80-84 RMS in micro inches or Ra value of 1.8 to 2.0 microns.