Learn More
TA Instruments Stainless Steel, Sandblasted, 40 mm, Quick Change Plate, HR X0, DHR, ARES-G2, APS
Catalog No. 5000580776 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments HR X0, DHR, ARES-G2, APS Stainless Steel Sandblasted Quick Change Plate (40 mm) features a surface finish of 80-84 RMS (Ra 1.8 to 2.0 microns). Compatible with Advanced Peltier Plates and ARES-G2 APS Standard Flat Plate for reliable placement and consistent measurements.
Catalog No. 50-005-80776 Supplier TA Instruments Supplier No. 402769.901
Waters TA Instruments HR X0, DHR, ARES-G2, APS Stainless Steel Sandblasted Quick Change Plate (40 mm) features a surface finish of 80-84 RMS (Ra 1.8 to 2.0 microns). Compatible with Advanced Peltier Plates and ARES-G2 APS Standard Flat Plate for reliable placement and consistent measurements.