Learn More
TA Instruments Stainless Steel, Sandblasted, 8 mm Plate, NST, HR X0
Catalog No. 5000580797 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments HR X0 Stainless Steel Sandblasted Upper Plate (8 mm, NST). Surface Finish: 80-84 RMS in micro inches or Ra value of 1.8 to 2.0 microns.
Catalog No. 50-005-80797 Supplier TA Instruments Supplier No. 519080.946
Waters TA Instruments HR X0 Stainless Steel Sandblasted Upper Plate (8 mm, NST). Surface Finish: 80-84 RMS in micro inches or Ra value of 1.8 to 2.0 microns.