Learn More
TA Instruments Stainless Steel, Sandblasted, 8 mm, Quick Change Plate, HR X0, DHR, ARES-G2, APS
Catalog No. 5000580778 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments HR X0, DHR, ARES-G2, APS Stainless Steel Sandblasted Quick Change Plate (8 mm) features a surface finish of 80-84 RMS (Ra 1.8 to 2.0 microns). Designed for Advanced Peltier Plates and ARES-G2 APS systems, it ensures secure placement, improved sample adhesion, and reproducible measurements.
Catalog No. 50-005-80778 Supplier TA Instruments Supplier No. 402811.901
Waters TA Instruments HR X0, DHR, ARES-G2, APS Stainless Steel Sandblasted Quick Change Plate (8 mm) features a surface finish of 80-84 RMS (Ra 1.8 to 2.0 microns). Designed for Advanced Peltier Plates and ARES-G2 APS systems, it ensures secure placement, improved sample adhesion, and reproducible measurements.