Learn More
Fisherbrand™ Stainless Steel Scalpel Blade, #10
Catalog No. 13812230 Shop All Fisherbrand Products
Change view
Click to view available options
Size:
# 10
Catalog No. Size
13-812-230 # 10
1 options
This item is not returnable. View return policy
Catalog No. 13-812-230
Add to Cart
Add to Cart
This item is not returnable. View return policy
These Stainless Steel Scalpel Blades, #10, are packaged in individual foil packs. For use in various applications.
  • Traditional curved cutting edge blade fits handles 3, 3L, 3 Graduated, 5B, 7, 9, B3 and B3L
  • Used for making small incisions in skin and muscle
  • Used in more specialized surgeries such as for harvesting radial artery during a coronary artery bypass operation, for opening a bronchus during thoracic surgery, and for inguinal hernia repair
Specifications
Blade Material Stainless Steel
For Use With (Application) Laboratory, Dissection, Dermaplaning
Size # 10
For Use With (Equipment) Surgical Design Scalpel Handle # 3
Disposable Single-use
Sterility Sterile
Certifications/Compliance ISO, GMP
Quantity 10/Pk.