Learn More
BURKLE INC Scoop AISI 304, V2A, capacity 0.66 gal. (2500 ml), Ø 5.91 in. (150 mm)
Catalog No. 5000025564
Encompass_Preferred
The stainless steel scoop can be used for taking samples of powders, granulates and pastes. Made of stainless steel AISI 304 (1.4301), the material is sterilizable and autoclavable. The rustproof stainless steel scoop has a smooth, high-gloss polished surface and is food-safe.
The tubular shape makes it easy to immerse the scoop in the medium being sampled. The high sides prevent the sample from trickling out. The round handle offers a good grip.
Catalog No. 50-000-25564 Supplier BURKLE INC Supplier No. 53702000
The stainless steel scoop can be used for taking samples of powders, granulates and pastes. Made of stainless steel AISI 304 (1.4301), the material is sterilizable and autoclavable. The rustproof stainless steel scoop has a smooth, high-gloss polished surface and is food-safe.
The tubular shape makes it easy to immerse the scoop in the medium being sampled. The high sides prevent the sample from trickling out. The round handle offers a good grip.