Learn More
BURKLE INC Stainless steel scoop, V2A, 45.3 in (115 cm), capacity 33.81 oz (1000 ml)
Catalog No. 5000026050
Encompass_Preferred
In contrast to conventional scoops, the stainless steel scoop has no openings, undercuts or crevices in which residues could collect. Pore-free finishing produced by high-quality laser welding seams.
The stainless steel scoop can be flame-sterilized and can therefore be made completely germ-free.
Catalog No. 50-000-26050 Supplier BURKLE INC Supplier No. 53541000
In contrast to conventional scoops, the stainless steel scoop has no openings, undercuts or crevices in which residues could collect. Pore-free finishing produced by high-quality laser welding seams.
The stainless steel scoop can be flame-sterilized and can therefore be made completely germ-free.