Learn More
Analytical Sales & Services Stainless Steel Screws
Catalog No. 5000806055
Encompass
18-8 Stainless Steel Button Head Hex Drive Screw\nUse to affix a Thermal Adapter Plate (TAP) to a Thermal Transfer Deck (TTD).\nTwo washers are required for each screw.\n\n• Length: 1"\n• Thread size: 6-32\n\nQty/Pkg: 100
Catalog No. 50-008-06055 Supplier Analytical Sales & Services Supplier No. TAPSCREW
18-8 Stainless Steel Button Head Hex Drive Screw\nUse to affix a Thermal Adapter Plate (TAP) to a Thermal Transfer Deck (TTD).\nTwo washers are required for each screw.\n\n• Length: 1"\n• Thread size: 6-32\n\nQty/Pkg: 100