Learn More
BINDER™ Rack
SDP
Catalog No. 08111883 Shop All BINDER Inc. Products
Change view
Click to view available options
Material:
Stainless Steel
Catalog No. Material
08-111-883 Stainless Steel
1 options
Catalog No. 08-111-883 Supplier BINDER™ Supplier No. 80122177
Add to Cart
Add to Cart
Material: Stainless Steel; For Use With: BF 260, FD 260, FED 260
  • Perforated
  • Manufactured from stainless steel for durability and dependability
Specifications
Description Shelf, perforated - Stainless steel
For Use With (Equipment) BINDER Chambers BF 260, FD 260, FED 260
Material Stainless Steel
Type Rack