Learn More
DWK Life Sciences Stainless Steel Shipping Bottle, UN certified, with DIN 168-1 Thread, GL 45
Shop All DWK Life Sciences Products

Description
- Certified to UN standards for the carriage of liquids classified as dangerous goods in UN Packing Groups I (high danger), II (medium danger), and III (low danger)
- No additional outer packaging required due to UN approval
- Suitable for liquids with a relative density of 2.0 or less
- Manufactured from corrosion-resistant AISI Type 316L (1.4404) stainless steel with all welded seams for hygienic construction
- Smooth inner surface finish corresponding to IIIc (DIN 17441) with rounded inner edges for ease of cleaning
- Polished and brushed durable exterior finish
- GL 45 thread with built-in pouring lip; supplied complete with GL 45 stainless steel screw cap and PTFE-surfaced platinum-catalysed silicone cap liner
- International regulations are subject to change; it is the user's responsibility to comply with all applicable laws and regulations
Specifications
Specifications
| Capacity (Metric) | 1500 mL |
| Color | Silver |
| For Use With (Application) | Carriage of liquids classified as dangerous goods in UN packing Groups I (high danager), II (medium danger) and III (low danger); Storage, transport and safe handling of hazardous, dangerous and valuable materials; Storage and transport of light-sensitive materials |
| Material | Stainless Steel (316L) |
| Nominal Capacity | 1500 mL |
| Closure Included | Yes |
| Capacity (English) | 51 oz. |
| Shape | Round |
| Product Type | Transport Bottle |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.