Learn More
TA Instruments Stainless Steel Single-Piece Concentric Cylinder Cup, Radius 15.2 mm, Compatible with Conical DIN, Recessed End, Vane, Wide Gap Vane Rotors, for HR X0, DHR and AR-Series Peltier and Electrically Heated Jacket
Catalog No. 5000580765 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Stainless Steel Single-Piece Concentric Cylinder Cup is for use with HR X0, DHR, and AR-Series Peltier Jacket systems as well as HR X0, DHR, and AR-Series Electrically Heated Jacket systems. The cup has a radius of 15.2 mm and is compatible with upper conical DIN, recessed end, vane, and wide gap vane rotors. Sample volume depends on rotor selection.
Catalog No. 50-005-80765 Supplier TA Instruments Supplier No. 547305.901
Waters TA Instruments Stainless Steel Single-Piece Concentric Cylinder Cup is for use with HR X0, DHR, and AR-Series Peltier Jacket systems as well as HR X0, DHR, and AR-Series Electrically Heated Jacket systems. The cup has a radius of 15.2 mm and is compatible with upper conical DIN, recessed end, vane, and wide gap vane rotors. Sample volume depends on rotor selection.