Learn More
Anton Paar STAINLESS STEEL, LUER ADAPTER, FOR PELTIER, 2.5mm for MCP 100
Supplier:  Anton Paar 143949
Encompass_Preferred
2.5 mm stainless steel sample cell (polarimeter tube) with wireless Toolmaster function which transfers temperature and path length to MCP 100.Filling volume approximately 0.05 mL.Connection to a syringe through Luer filling ports.Suitable for use with Anton Paar polarimeter MCP 100 item NC1818306
Catalog No. NC1818313