Learn More
Electron Microscopy Sciences Stainless Steel Spatula #4
SDP
Catalog No. 50996778
Encompass_Preferred
Made from polished stainless steel. They are ideal for applying wax to glass knives, colloidal adhesives to stubs, and for mixing embedding resins.
Catalog No. 50-996-778 Supplier Electron Microscopy Sciences Supplier No. 6212714
Made from polished stainless steel. They are ideal for applying wax to glass knives, colloidal adhesives to stubs, and for mixing embedding resins.