Learn More
Electron Microscopy Sciences Stainless Steel Spatulas Kit include Four Spatulas
SDP
Catalog No. 501930973
Encompass_Preferred
Spatulas are general purpose tools for scraping, getting into small areas, working with adhesives, and moving moving small electronic parts. Made of rust-free stainless steel AISI 410 with serrated handles for a better grip. Includes these four styles: 78326-12, 78326-13, 78326-14, 78326-15.
Catalog No. 50-193-0973 Supplier Electron Microscopy Sciences Supplier No. 7832592
Spatulas are general purpose tools for scraping, getting into small areas, working with adhesives, and moving moving small electronic parts. Made of rust-free stainless steel AISI 410 with serrated handles for a better grip. Includes these four styles: 78326-12, 78326-13, 78326-14, 78326-15.