Learn More
Electron Microscopy Sciences Stainless Steel Specimen Basket
SDP
Catalog No. 5028797
Encompass_Preferred
Both the top and the bottom are made from a stainless steel mesh, which offers good fluid circulation. Has a male and a female threading for secure closure.
Catalog No. 50-287-97 Supplier Electron Microscopy Sciences Supplier No. 7019001
Both the top and the bottom are made from a stainless steel mesh, which offers good fluid circulation. Has a male and a female threading for secure closure.