Learn More
Electron Microscopy Sciences Stainless Steel Specimen Basket
SDP
Supplier:  Electron Microscopy Sciences 7019006
Encompass_Preferred
Both the top and the bottom are made from a stainless steel mesh, which offers good fluid circulation. Has a male and a female threading for secure closure.
Catalog No. 50-287-96