Learn More
Med Vet International Stainless Steel Spike Washer, for use with 2.7mm screw, J0375WC
Catalog No. 5000582612
Encompass_Preferred
Used for ligament attachment or fixation to spread load or retain wire or sutures. Autoclavable.
Catalog No. 50-005-82612 Supplier Med Vet International Supplier No. JOR375WC
Used for ligament attachment or fixation to spread load or retain wire or sutures. Autoclavable.