Learn More
Med Vet International Stainless Steel Spike Washer, for use with 3.5mm screw, J0375WD
Catalog No. 5000582696
Encompass_Preferred
Used for ligament attachment or fixation to spread load or retain wire or sutures. Autoclavable.
Catalog No. 50-005-82696 Supplier Med Vet International Supplier No. JOR375WD
Used for ligament attachment or fixation to spread load or retain wire or sutures. Autoclavable.