Learn More
TA Instruments Stainless Steel Standard Concentric Cylinder Cup with Cap, Radius 15.2 mm, Two-Piece Screw-Off Bottom, Compatible with Upper Conical DIN, Recessed End, Vane, Wide Gap Vane Rotors, for HR X0, DHR and AR-Series Peltier and Electrically Heated Jacket
Catalog No. 5000580701 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Stainless Steel Standard Concentric Cylinder Cup With Cap is for use with HR X0, DHR, and AR-Series Peltier Jacket and HR X0, DHR, and AR-Series Electrically Heated Jacket. Radius is 15.2 mm. It features a convenient two-piece design with screw-off bottom that simplifies sample cleaning. Compatible with upper conical DIN, recessed end, vane, and wide gap vane rotors. Sample volume depends on rotor selection.
Catalog No. 50-005-80701 Supplier TA Instruments Supplier No. 545696.901
Waters TA Instruments Stainless Steel Standard Concentric Cylinder Cup With Cap is for use with HR X0, DHR, and AR-Series Peltier Jacket and HR X0, DHR, and AR-Series Electrically Heated Jacket. Radius is 15.2 mm. It features a convenient two-piece design with screw-off bottom that simplifies sample cleaning. Compatible with upper conical DIN, recessed end, vane, and wide gap vane rotors. Sample volume depends on rotor selection.