Learn More
TA Instruments Stainless Steel Standard Concentric Cylinder Cup, Sandblasted, Radius 15.2 mm, Compatible with Upper Conical DIN, Recessed End, Vane, Wide Gap Vane Rotors, Surface Finish 80-84 RMS, Ra 1.8-2.0 Microns
Catalog No. 5000580882 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Stainless Steel Standard Concentric Cylinder Cup, Sandblasted, has a radius of 15.2 mm. Compatible with upper conical DIN, recessed end, vane, and wide gap vane rotors. Sample volume depends on rotor selection. Surface finish is 80-84 RMS in micro inches or Ra value of 1.8 to 2.0 microns.
Catalog No. 50-005-80882 Supplier TA Instruments Supplier No. 546046.901
Waters TA Instruments Stainless Steel Standard Concentric Cylinder Cup, Sandblasted, has a radius of 15.2 mm. Compatible with upper conical DIN, recessed end, vane, and wide gap vane rotors. Sample volume depends on rotor selection. Surface finish is 80-84 RMS in micro inches or Ra value of 1.8 to 2.0 microns.