Learn More
Alkali Scientific ASI™ Stainless Steel Stap back Cassette, 5 x 7 (13x18cm) Stainless Steel frame & crossbars. Aluminum Door. Abrasion resistant plastic front. Thick, non-shedding, oil free felt interior to assure smooth film contact